Learn More
Gold Biotechnology Inc Nickel NTA Agarose Beads 25 mL
SDP
Catalog No. 501532798
Encompass_Preferred
Nickel-NTA Agarose Resin has a binding capacity of ~50 mg/ml. In addition, it allows for purification of proteins under native or denaturing conditions. GoldBio Nickel Agarose works very well with the His-Tag Buffer Set.
Catalog No. 50-153-2798 Supplier Gold Biotechnology Inc Supplier No. H35025
Nickel-NTA Agarose Resin has a binding capacity of ~50 mg/ml. In addition, it allows for purification of proteins under native or denaturing conditions. GoldBio Nickel Agarose works very well with the His-Tag Buffer Set.