Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nickel Peroxide

Catalog No. 722625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-262-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 72-262-5G Supplier Sigma Aldrich Supplier No. 722625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst for: One-dimensional oxide-metal hybrid structures and site-specific enhanced reactivity for CO oxidationMicrowave induced catalytic degradation of crystal violetUsed to study differences in reactivity trends for carbon monoxide catalyzed oxidation caused by anionic transition metal oxide clustersReagent: For solvent-free oxidation of alcohols†For chemoselective oxidation of organic compounds using inductive heating To improve the synthesis of methoxy cephalosporin'sas intermediateOxidant for the transformation of alcohols into acids and hydrazones into diazoalkanes

Specifications

CAS 7440-48-4
Density 8.9 g/mL at 25°C
Boiling Point 2900°C
Molecular Formula Co
Linear Formula Co
Quantity 5 g
MDL Number MFCD00010935
Synonym Nickel dioxide; Nickel oxide
Molecular Weight (g/mol) 58.93 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.