Learn More
Cole-Parmer Cole-Parmer Regular Holder, Nickel-Plated, 0 to 0.71" Dia Rod s
Catalog No. NC1534644 Shop All Cole-Parmer Products
Encompass_Preferred
Ideal for holding clamps to lab-frames, at a 90 angle.
Catalog No. NC1534644 Supplier Cole-Parmer Supplier No. 0804120
Ideal for holding clamps to lab-frames, at a 90 angle.