Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel plating solution, electroless, ammonia free

Catalog No. AA44070AK
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44070AK 250 mL
AA44070K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA44070AK Supplier Thermo Scientific Chemicals Supplier No. 044070AK

For plating on silicon, germainium, gallium arsenide, cadmium sulfide, nickel or Kovar

For plating on silicon, germainium, gallium arsenide, cadmium sulfide, nickel or Kovar

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Chemical Name or Material Nickel plating solution
Name Note Electroless, ammonia free
Quantity 250 mL
MDL Number MFCD00011137
UN Number UN3082

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.