Learn More
Glass Expansion Nickel Sampler Cone with Copper Base for Shimadzu 2030 ICP-MS
SDP
Supplier:  Glass Expansion SZ7001NI/CU
Encompass_Preferred
Nickel Sampler Cone with Copper Base ¿ Standard sampler cone for Shimadzu 2030 ICP-MS. Suitable for most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic) and provides better heat ransfer. Not recommended for samples containing HF. Use a new Graphite Sampler Cone Gasket (SZ5001) each time the cone is re-installed, even if it¿s the same cone. Replace sampler cone screws if corroded or worn (SZ5002). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For standard nickel sampler cone for use with routine samples, see catalog number 501907696.For standard low-cost copper sampler cone see 501907695.For corrosive samples, including HF, use Platinum Sampler Cones, see catalog numbers 501907701 (15 mm Pt insert), 501907702 (10 mm Pt insert).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7697