Learn More
Glass Expansion Nickel Skimmer Cone for Thermo Scientific Element 1/Element 2/Neptune/Neoma ICP-MS (1067600)
SDP
Catalog No. 501907706
Encompass_Preferred
Nickel Skimmer Cone (1067600) ¿ Suitable for Thermo/Finnigan Neoma MC-ICP-MS, Element 1, 2 & Neptune. For lowest copper backgrounds. Not recommended for samples containing hydrofluoric acid (HF). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For lowest boron background, semiconductor applications, and samples containing HF along with other highly corrosive samples where acid concentration begin to exceed 5%, we recommend a platinum cone, see catalog number 501907714.For high sensitivity applications, we recommend 501907707.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7706 Supplier Glass Expansion Supplier No. TF1002ANI
Nickel Skimmer Cone (1067600) ¿ Suitable for Thermo/Finnigan Neoma MC-ICP-MS, Element 1, 2 & Neptune. For lowest copper backgrounds. Not recommended for samples containing hydrofluoric acid (HF). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For lowest boron background, semiconductor applications, and samples containing HF along with other highly corrosive samples where acid concentration begin to exceed 5%, we recommend a platinum cone, see catalog number 501907714.For high sensitivity applications, we recommend 501907707.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001