Learn More
Glass Expansion Nickel Sampler Cone with Copper Core for Thermo Scientific Element 1/Element 2/Neptune/Neoma ICP-MS (1044530)
SDP
Catalog No. 501907705
Encompass_Preferred
Nickel Sampler Cone with Copper Core (1044530) ¿ Standard sampler cone for Thermo/Finnigan Neoma MC-ICP-MS and Element 1, 2 & Neptune. Suitable for most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic). Not recommended for samples containing hydrofluoric acid (HF). Use a new Graphite Sampler Cone Gasket (501907717) each time the cone is re-installed, even if it¿s the same cone. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For lowest boron background and semiconductor applications, we recommend a platinum cone, see catalog number 501907711.For high sensitivity applications and low matrix containing samples, we reccommend Nickel Jet Sampler, see catalog number 501907709.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7705 Supplier Glass Expansion Supplier No. TF1001NI/CU
Nickel Sampler Cone with Copper Core (1044530) ¿ Standard sampler cone for Thermo/Finnigan Neoma MC-ICP-MS and Element 1, 2 & Neptune. Suitable for most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic). Not recommended for samples containing hydrofluoric acid (HF). Use a new Graphite Sampler Cone Gasket (501907717) each time the cone is re-installed, even if it¿s the same cone. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For lowest boron background and semiconductor applications, we recommend a platinum cone, see catalog number 501907711.For high sensitivity applications and low matrix containing samples, we reccommend Nickel Jet Sampler, see catalog number 501907709.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001