Learn More
Glass Expansion Nickel Skimmer Cone, Xi, Xt, Copper Core for for Thermo Scientific X Series ICP-MS (3600811)
SDP
Catalog No. 501907723
Encompass_Preferred
Nickel Micro-Skimmer Cone, Xi, Xt, Copper Core (3600811) ¿ Suitable for Thermo/VG X-Series ICP-MS. Provides high matrix tolerance. Ideal for applications with high total dissolved solids (TDS). Not recommended for samples containing hydrofluoric acid (HF). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1028) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For samples containing HF, and other highly corrosive samples where acid concentration begin to exceed 5%, we recommend a platinum cone, see catalog number 501907721.For high performance and sensitivity, we recommend 501907719.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7723 Supplier Glass Expansion Supplier No. TG1022NI
Nickel Micro-Skimmer Cone, Xi, Xt, Copper Core (3600811) ¿ Suitable for Thermo/VG X-Series ICP-MS. Provides high matrix tolerance. Ideal for applications with high total dissolved solids (TDS). Not recommended for samples containing hydrofluoric acid (HF). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1028) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For samples containing HF, and other highly corrosive samples where acid concentration begin to exceed 5%, we recommend a platinum cone, see catalog number 501907721.For high performance and sensitivity, we recommend 501907719.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001