Learn More
Glass Expansion Nickel Skimmer Cone - 7500c, Suitable for: Agilent 7500c (G1820-65497)
SDP
Catalog No. 501907565
Encompass_Preferred
Nickel Skimmer Cone (G1820-65497). For Agilent 4500/7500c/7500cs ICP-MS. Standard cone used with 7500c. Suitable for most common applications (e.g., routine aqueous environmental samples, <5% acid matrix, non-HF). Not recommended for samples containing hydrofluoric acid (HF). Use with Nickel sampler cone. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For samples containing HF, and other highly corrosive samples, we recommend a platinum sampler cone, see catalog number 501907569.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7565 Supplier Glass Expansion Supplier No. AT1002CNI
Nickel Skimmer Cone (G1820-65497). For Agilent 4500/7500c/7500cs ICP-MS. Standard cone used with 7500c. Suitable for most common applications (e.g., routine aqueous environmental samples, <5% acid matrix, non-HF). Not recommended for samples containing hydrofluoric acid (HF). Use with Nickel sampler cone. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For samples containing HF, and other highly corrosive samples, we recommend a platinum sampler cone, see catalog number 501907569.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001