Learn More
Sigma Aldrich Nickel on silica/alumina
Catalog No. 208779100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
20-877-9100G 100 g
1 options
Catalog No. 20-877-9100G Supplier Sigma Aldrich Supplier No. 208779100G
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Nickel on silica/alumina can be used as an efficient catalyst in a variety of applications such as: in plastic waste pyrolysis for the production of fuelto study the supercritical water reforming of glycerolin the supercritical water reforming of acetic acid for the production of hydrogenWe are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.
Specifications
MDL Number MFCD00011137
Quantity 100 g