Learn More
Glass Expansion Nickel Skimmer Cone for x-lens Agilent 7900/8900 ICP-MS (G8400-67200)
SDP
Catalog No. 501907593
Encompass_Preferred
Nickel Skimmer Cone for x-lens (G8400-67200) ¿ For Agilent 7900/8900 ICP-MS. Standard cone used with x-lens and suitable for most common applications (e.g., routine aqueous environmental samples, <5% acid matrix, non-HF, non-organic). Not for samples containing hydrofluoric acid (HF). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For corrosive acids, including HF, use Pt Skimmer with Cu Base for x-lens, P/N 501907594, must be mounted on brass adaptor skimmer base. For organic analysis, use Pt Skimmer with Ni Base for x-lens, P/N 501907595, which must be mounted on brass adaptor skimmer base.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7593 Supplier Glass Expansion Supplier No. AT7902XNI
Nickel Skimmer Cone for x-lens (G8400-67200) ¿ For Agilent 7900/8900 ICP-MS. Standard cone used with x-lens and suitable for most common applications (e.g., routine aqueous environmental samples, <5% acid matrix, non-HF, non-organic). Not for samples containing hydrofluoric acid (HF). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For corrosive acids, including HF, use Pt Skimmer with Cu Base for x-lens, P/N 501907594, must be mounted on brass adaptor skimmer base. For organic analysis, use Pt Skimmer with Ni Base for x-lens, P/N 501907595, which must be mounted on brass adaptor skimmer base.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001