Learn More
Elemental Scientific Inc Nickel Skimmer Cone, Insert Ready for Thermo ICPMS
SDP
Catalog No. NC2828619
Encompass_Preferred
Nickel skimmer cone with mount for insert (not included). Requires use of one of three inserts - 2.8mm High Sensitivity (ES-3000-1236), 3.5mm High Matrix (ES-3000-1230-T), or 4.5mm Robust (MC-06591). For Thermo Scientific iCAP Q/RQ/TQ models.
Catalog No. NC2828619 Supplier Elemental Scientific Inc Supplier No. ES30001229T
Nickel skimmer cone with mount for insert (not included). Requires use of one of three inserts - 2.8mm High Sensitivity (ES-3000-1236), 3.5mm High Matrix (ES-3000-1230-T), or 4.5mm Robust (MC-06591). For Thermo Scientific iCAP Q/RQ/TQ models.