Learn More
Glass Expansion Nickel Skimmer Cone for PerkinElmer NexION 5000, an economic alternative to platinum (N8161117)
SDP
Catalog No. 502702952
Encompass_Preferred
Nickel Skimmer Cone ¿ Nickel skimmer cone for PerkinElmer NexION 5000 ICP-MS. An economic alternative to platinum and suitable for most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic). Not recommended for samples containing HF. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For corrosive samples, including HF, we recommend Platinum Skimmer Cones, see catalog number PE4014-Pt.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2952 Supplier Glass Expansion Supplier No. PE4012NI
Nickel Skimmer Cone ¿ Nickel skimmer cone for PerkinElmer NexION 5000 ICP-MS. An economic alternative to platinum and suitable for most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic). Not recommended for samples containing HF. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For corrosive samples, including HF, we recommend Platinum Skimmer Cones, see catalog number PE4014-Pt.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001