Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Standard, Inorganic, 1,000 μg/mL, 125 mL

Catalog No. USICP028
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-ICP-028 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-ICP-028 Supplier Agilent Technologies Supplier No. ICP028
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7440-02-0 , 7697-37-2 , 7732-18-5
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

CAS Max % 97.9, 2.0, 0.1
Analyte or Component Names Water,Nitric acid,Nickel
Element Water, Nitric Acid, Ni
Solvent or Matrix Water/Dilute Nitric Acid
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Nickel Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.