Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel standard solution, for AAS, 1 mg/ml Ni in 2-5% HNO3

Catalog No. AC196151000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196151000 100 mL
AC196155000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC196151000 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 196151000
Only null left
Add to Cart

CAS: 7440-02-0 | Ni | 58.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nickel standard solution
Name Note 1 mg/ml Ni in 2% HNO3
Packaging Plastic bottle
CAS 7440-02-0, 13478-00-7, 7697-37-2, 7732-18-5
CAS Min % 1.9, 96.9
CAS Max % 2.1, 97.1
Density 1.0320g/mL
Specific Gravity 1.032
Physical Form Crystals, Crystalline Powder and/or Chunks
Molecular Formula Ni
Concentration 990 to 1010 μg/mL Ni in 2 to 5% HNO3 (20°C)
MDL Number MFCD00011137 MFCD06798735
Color Green
Solubility Soluble in water
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES [Ni]
IUPAC Name nickel
Molecular Weight (g/mol) 58.69
Quantity 100 mL
PubChem CID 935
ChEBI CHEBI:28112
Formula Weight 58.69
Grade AAS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.