Learn More
Hach Company Nickel Standard Solution, 1000 mg/L as Ni (NIST), 100 mL
Catalog No. 501994944 Shop All Hach Company Products
Encompass
For accuracy checks in nickel determinations. (HNO3 matrix)
Catalog No. 50-199-4944 Supplier Hach Company Supplier No. 1417642
For accuracy checks in nickel determinations. (HNO3 matrix)