Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Sulfate ACS AR Powder, Macron™

Catalog No. MK6400212 Shop All Avantor Products
Change view
Click to view available options
STAKMOR; 2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-6400-212 STAKMOR; 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-6400-212 Supplier Avantor Supplier No. 640005

missing translation for 'validMainframeMsg'

Specifications

CAS 10101-97-0
Physical Form Powder
Quantity 2.5 kg
Molecular Formula H12NiO10S
Synonym nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
IUPAC Name nickel(2+);sulfate;hexahydrate
Molecular Weight (g/mol) 262.839
PubChem CID 5284429
ChEBI CHEBI:53437
Formula Weight 262.84
Grade ACS
Packaging STAKMOR
Chemical Name or Material Nickel Sulfate, 6-Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.