Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Test Solutions, LaMotte™
SDPSCI_ED

Catalog No. S95016 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S95016 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95016 Supplier LaMotte Company Supplier No. 366301SC/DEL

  • Hydrochloric Acid 2.5N (60mL)
  • Ammonium Persulfate (30g)
  • Silver Nitrate 0.0141N (30mL)
  • Sodium Citrate 10% (250mL)
  • Dimethylglyoxime 1% (60mL)
  • Ammonium Hyudroxide Concentrate (60mL)
  • 3 drippers, spoon, test tube and graduated cylander

Specifications

Quantity 1 Kit
Chemical Name or Material Nickel Test Solutions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.