Learn More
Perkin Elmer US LLC Nickel Reference Material - Wire, 780°C
Catalog No. 509044716
Encompass_Preferred
PerkinElmer’s Calibration and Reference Standards ensure accurate thermogravimetric measurements for all of PerkinElmer’s TGA instruments. Magnetic calibration and reference standards are available as a kit, as well as individually. PerkinElmer recommends the use of a fresh reference material every time you calibrate your instruments. This helps to avoid the effects of hysteresis and oxidation on the Curie Point of the reference materials.
Catalog No. 50-904-4716 Supplier Perkin Elmer US LLC Supplier No. N5190869
PerkinElmer’s Calibration and Reference Standards ensure accurate thermogravimetric measurements for all of PerkinElmer’s TGA instruments. Magnetic calibration and reference standards are available as a kit, as well as individually. PerkinElmer recommends the use of a fresh reference material every time you calibrate your instruments. This helps to avoid the effects of hysteresis and oxidation on the Curie Point of the reference materials.