Learn More
Cayman Chemical Niclosamide, 50-65-7, 25 g
SDP
Supplier:  Cayman Chemical 1064925
Encompass_Preferred
Molecular Formula C13H8Cl2N2O4, Purity ≥95%, Formula Weight 327.1, Synonyms: NSC 178296; A salicylanilide compound with antihelminthic actions; specifically inhibits STAT3 (IC50 = 0.25 M); inhibits the proliferation of Du145 prostate cancer cells, which have constitutively active STAT3 (IC50 = 0.7 M).
Catalog No. NC0754158