Learn More
Medchemexpress LLC Niclosamide (olamine) | 1420-04-8 | 98.7% | 1 G
SDP
Catalog No. 5000409454
Encompass_Preferred
Niclosamide olamine is an orally active antihelminthic agent used in parasitic infection research. It functions as a STAT3 inhibitor with an IC50 of 0.25 μM in HeLa cells. This compound also exhibits biological activities against cancer and inhibits DNA replication in Vero E6 cells.
  • Inhibits adrenocortical carcinoma cellular proliferation.
  • Inhibits STAT3-mediated luciferase reporter activity in HeLa cells.
  • Inhibits virus replication in Vero E6 cells.
  • Inhibits adrenocortical carcinoma tumor growth in vivo.
Catalog No. 50-004-09454 Supplier Medchemexpress LLC Supplier No. HYB0497C1G
Niclosamide olamine is an orally active antihelminthic agent used in parasitic infection research. It functions as a STAT3 inhibitor with an IC50 of 0.25 μM in HeLa cells. This compound also exhibits biological activities against cancer and inhibits DNA replication in Vero E6 cells.
  • Inhibits adrenocortical carcinoma cellular proliferation.
  • Inhibits STAT3-mediated luciferase reporter activity in HeLa cells.
  • Inhibits virus replication in Vero E6 cells.
  • Inhibits adrenocortical carcinoma tumor growth in vivo.