Learn More
Medchemexpress LLC Niclosamide olamine | 1420-04-8 | 98.7% | 1 ML
SDP
Catalog No. 5000406837
Encompass_Preferred
Niclosamide olamine is an orally active antihelminthic agent primarily used in parasitic infection research. It functions as a STAT3 inhibitor with an IC50 of 0.25 μM in HeLa cells. This compound also demonstrates biological activities against various cancers and inhibits DNA replication in Vero E6 cells.
  • Orally active antihelminthic agent
  • Used in parasitic infection research
  • STAT3 inhibitor (IC50 of 0.25 μM in HeLa cells)
  • Exhibits biological activities against cancer
  • Inhibits DNA replication in Vero E6 cells
  • Suppresses adrenocortical carcinoma cellular proliferation
  • Reduces cyclin D1 and vimentin protein expression in human endometrial cancer cells
  • Inhibits metabolic viability of AN3CA, Hec1B, and Ishikawa cells
  • Suppresses colony formation in endometrial cancer cells
  • Inhibits invasive properties of endometrial cancer cells
Catalog No. 50-004-06837 Supplier Medchemexpress LLC Supplier No. HYB0497C10MM/1ML
Niclosamide olamine is an orally active antihelminthic agent primarily used in parasitic infection research. It functions as a STAT3 inhibitor with an IC50 of 0.25 μM in HeLa cells. This compound also demonstrates biological activities against various cancers and inhibits DNA replication in Vero E6 cells.
  • Orally active antihelminthic agent
  • Used in parasitic infection research
  • STAT3 inhibitor (IC50 of 0.25 μM in HeLa cells)
  • Exhibits biological activities against cancer
  • Inhibits DNA replication in Vero E6 cells
  • Suppresses adrenocortical carcinoma cellular proliferation
  • Reduces cyclin D1 and vimentin protein expression in human endometrial cancer cells
  • Inhibits metabolic viability of AN3CA, Hec1B, and Ishikawa cells
  • Suppresses colony formation in endometrial cancer cells
  • Inhibits invasive properties of endometrial cancer cells