Learn More
Medchemexpress LLC Niclosamide (olamine) | 1420-04-8 | 98.7% | 5 G
SDP
Catalog No. 5000407101
Encompass_Preferred
Niclosamide olamine (BAY2353) is an orally active antihelminthic agent used in parasitic infection research. It is a STAT3 inhibitor with an IC50 of 0.25 μM in HeLa cells. Niclosamide olamine has biological activities against cancer and inhibits DNA replication in Vero E6 cells.
  • Functions as an orally active antihelminthic agent in parasitic infection research.
  • Acts as a STAT3 inhibitor with an IC50 of 0.25 μM in HeLa cells.
  • Shows biological activity against cancer.
  • Inhibits DNA replication in Vero E6 cells.
  • Inhibits adrenocortical carcinoma cellular proliferation in BD140A, SW-13, and NCI-H295R cells.
  • Inhibits STAT3-mediated luciferase reporter activity in HeLa cells.
  • Inhibits virus replication in Vero E6 cells.
Catalog No. 50-004-07101 Supplier Medchemexpress LLC Supplier No. HYB0497C5G
Niclosamide olamine (BAY2353) is an orally active antihelminthic agent used in parasitic infection research. It is a STAT3 inhibitor with an IC50 of 0.25 μM in HeLa cells. Niclosamide olamine has biological activities against cancer and inhibits DNA replication in Vero E6 cells.
  • Functions as an orally active antihelminthic agent in parasitic infection research.
  • Acts as a STAT3 inhibitor with an IC50 of 0.25 μM in HeLa cells.
  • Shows biological activity against cancer.
  • Inhibits DNA replication in Vero E6 cells.
  • Inhibits adrenocortical carcinoma cellular proliferation in BD140A, SW-13, and NCI-H295R cells.
  • Inhibits STAT3-mediated luciferase reporter activity in HeLa cells.
  • Inhibits virus replication in Vero E6 cells.