Learn More
AdipoGen Niclosamide Ethanolamine, 1420-04-8, MFCD01941566, 25 mg
SDP
Supplier:  AdipoGen AGCR13644M025
Encompass_Preferred
MF: C13H8Cl2N2O4 . C2H7NO, MW: 388.206, ≥98%, Appearance: Yellow to brown solid. Antihelminthic, molluscicide and trypanocidal agent that inhibits mitochondrial oxidative phosphorylation and anaerobic ATP production. Anticancer agent with antiproliferative activity in a broad spectrum of cancer cells. Targets and inhibits multiple signaling pathways, including STAT3, NF-kB, Wnt/beta-catenin, Notch and mTORC1.
Catalog No. 50-117-6142