Learn More
Cayman Chemical Niclosamide (ethanolamine salt), Purity: ≥98%, CAS: 1420-04-8, 100 mg
SDP
Catalog No. 502965307
Encompass_Preferred
A molluscicide; toxic to H. trivolvis snail eggs, juveniles, and adults (LC50s = 0.035, 0.044, and 0.11 mg/kg, respectively), as well as rainbow trout, white sucker, and fathead minnow (LC50s = 0.03, 0.09, and 0.11 mg/L, respectively); induces cell cycle arrest at the G0/G1 phase and induces apoptosis in DU145 cells in a concentration-dependent manner; inhibits STAT3-induced gene expression in a reporter assay using HeLa cells at 5 µM; uncouples mitochondria from respiration and increases the oxygen consumption rate of NIH3T3 fibroblasts at 1 µM; reduces blood glucose and plasma insulin levels, as well as decreases liver weight and triglyceride levels, in a mouse model of high-fat diet-induced diabetes in the diet at 1,500 ppm
Catalog No. 50-296-5307 Supplier Cayman Chemical Supplier No. 17118100 MG
A molluscicide; toxic to H. trivolvis snail eggs, juveniles, and adults (LC50s = 0.035, 0.044, and 0.11 mg/kg, respectively), as well as rainbow trout, white sucker, and fathead minnow (LC50s = 0.03, 0.09, and 0.11 mg/L, respectively); induces cell cycle arrest at the G0/G1 phase and induces apoptosis in DU145 cells in a concentration-dependent manner; inhibits STAT3-induced gene expression in a reporter assay using HeLa cells at 5 µM; uncouples mitochondria from respiration and increases the oxygen consumption rate of NIH3T3 fibroblasts at 1 µM; reduces blood glucose and plasma insulin levels, as well as decreases liver weight and triglyceride levels, in a mouse model of high-fat diet-induced diabetes in the diet at 1,500 ppm