Learn More
Aviva Systems Biology Corp NICN1 Antibody - middle region (ARP60478_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60478_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NICN1. It was validated on Western Blot. . Alternate names: MGC12936ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-058