Learn More
Cayman Chemical NicoinamID MononucleoID 100mg
SDP
Catalog No. 502970244
Encompass_Preferred
An intermediate in the biosynthesis of NAD+ that has been used to enhance NAD biosynthesis and glucose-stimulated insulin secretion in Nampt+/- mice and high-fat diet-induced type 2 diabetes mice
Catalog No. 50-297-0244 Supplier Cayman Chemical Supplier No. 16411100 MG
An intermediate in the biosynthesis of NAD+ that has been used to enhance NAD biosynthesis and glucose-stimulated insulin secretion in Nampt+/- mice and high-fat diet-induced type 2 diabetes mice