Learn More
Thermo Scientific Nicolet Is50 Desiccant And Hum
Supplier:  Thermo Scientific 714081800
Encompass
desiccant and humidity indicator card absorbs moisture, changes color from blue to pink at relative humidity threshold for monitoring humidity levels inside scientific instrument housing to protect sensitive components from excess moisture.
Catalog No. 50-134-8811