Learn More
Selleck Chemical LLC Nicorandil S1971-50mg
SDP
Catalog No. 503110611
Encompass_Preferred
Nicorandil (SG-75) is a potassium channel activator and stimulates guanylate cyclase to increase formation of cyclic GMP (cGMP)
Catalog No. 50-311-0611 Supplier Selleck Chemical LLC Supplier No. S197150MG
Nicorandil (SG-75) is a potassium channel activator and stimulates guanylate cyclase to increase formation of cyclic GMP (cGMP)