Learn More
Medchemexpress LLC Nicorandil-d4 | 1132681-23-2 | 99.8% | 1 MG
SDP
Catalog No. 5000406202
Encompass_Preferred
Nicorandil-d4 (SG-75-d4) is the deuterium-labeled Nicorandil. Nicorandil (SG-75) is a potent potassium channel activator that targets vascular nucleoside diphosphate-dependent K+ channels and cardiac ATP-sensitive K+ channels (KATP). It is a nicotinamide ester with vasodilatory and cardioprotective effects, with potential use for angina and ischemic heart diseases. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Targets vascular nucleoside diphosphate-dependent K+ channels and cardiac ATP-sensitive K+ channels (KATP).
  • Has vasodilatory and cardioprotective effects.
  • Potential for angina and ischemic heart diseases.
Catalog No. 50-004-06202 Supplier Medchemexpress LLC Supplier No. HYB0341S1MG
Nicorandil-d4 (SG-75-d4) is the deuterium-labeled Nicorandil. Nicorandil (SG-75) is a potent potassium channel activator that targets vascular nucleoside diphosphate-dependent K+ channels and cardiac ATP-sensitive K+ channels (KATP). It is a nicotinamide ester with vasodilatory and cardioprotective effects, with potential use for angina and ischemic heart diseases. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Targets vascular nucleoside diphosphate-dependent K+ channels and cardiac ATP-sensitive K+ channels (KATP).
  • Has vasodilatory and cardioprotective effects.
  • Potential for angina and ischemic heart diseases.