Learn More
Medchemexpress LLC Nicorandil-d4 | 1132681-23-2 | 99.8% | 5 MG
SDP
Catalog No. 5000406212
Encompass_Preferred
Nicorandil-d4 is the deuterium-labeled form of Nicorandil, a potent potassium channel activator. It targets vascular nucleoside diphosphate-dependent K+ channels and cardiac ATP-sensitive K+ channels (KATP), exhibiting vasodilatory and cardioprotective effects. This compound has potential applications for angina and ischemic heart diseases.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Storage (Powder): -20°C for 3 years; 4°C for 2 years
  • Storage (In solvent): -80°C for 6 months; -20°C for 1 month
Catalog No. 50-004-06212 Supplier Medchemexpress LLC Supplier No. HYB0341S5MG
Nicorandil-d4 is the deuterium-labeled form of Nicorandil, a potent potassium channel activator. It targets vascular nucleoside diphosphate-dependent K+ channels and cardiac ATP-sensitive K+ channels (KATP), exhibiting vasodilatory and cardioprotective effects. This compound has potential applications for angina and ischemic heart diseases.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Storage (Powder): -20°C for 3 years; 4°C for 2 years
  • Storage (In solvent): -80°C for 6 months; -20°C for 1 month