Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicorandil 98.0+%, TCI America™
SDP

Catalog No. N08371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08371G 1 g
N0837100MG 100 mg
N08375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N08371G Supplier TCI America Supplier No. N08371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65141-46-0
Molecular Formula C8H9N3O4
Molecular Weight (g/mol) 211.18
MDL Number MFCD00186520
InChI Key LBHIOVVIQHSOQN-UHFFFAOYSA-N
Synonym nicorandil, ikorel, 2-nicotinamidoethyl nitrate, adancor, dancor, nicorandilum, sigmart, 2-pyridine-3-carbonylamino ethyl nitrate, n-2-hydroxyethyl nicotinamide nitrate, nicorandilum inn-latin
PubChem CID 47528
ChEBI CHEBI:31905
IUPAC Name 2-[(pyridin-3-yl)formamido]ethyl nitrate
SMILES [O-][N+](=O)OCCNC(=O)C1=CC=CN=C1

Specifications

Melting Point 90°C
Color White
Quantity 1 g
Formula Weight 211.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nicorandil
RTECSNumber US4667600
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.