Learn More
Cayman Chemical Nicorandil-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1132681-23-2, 1 mg
SDP
Catalog No. 502949887
Encompass_Preferred
An internal standard for the quantification of nicorandil by GC- or LC-MS
Catalog No. 50-294-9887 Supplier Cayman Chemical Supplier No. 336081 MG
An internal standard for the quantification of nicorandil by GC- or LC-MS