Learn More
Medchemexpress LLC Nicosulfuron-d6 | 1189419-41-7 | 98.4% | 1 MG
SDP
Catalog No. 5000412396
Encompass_Preferred
Nicosulfuron-d6 is a deuterium-labeled version of Nicosulfuron, a selective herbicide from the sulfonylurea family. It is commonly applied as a post-emergence herbicide to safeguard maize crops from weeds by inhibiting the activity of the acetolactate synthase (ALS) enzyme.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can impact the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-12396 Supplier Medchemexpress LLC Supplier No. HYB1876S1MG
Add to Cart
Add to Cart
Nicosulfuron-d6 is a deuterium-labeled version of Nicosulfuron, a selective herbicide from the sulfonylurea family. It is commonly applied as a post-emergence herbicide to safeguard maize crops from weeds by inhibiting the activity of the acetolactate synthase (ALS) enzyme.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can impact the pharmacokinetic and metabolic profiles of drugs.