Learn More
Medchemexpress LLC HY-N1475 5mg Medchemexpress, Nicotiflorin CAS:17650-84-9 Purity:>98%
SDP
Catalog No. 501970345
Encompass_Preferred
Medchemexpress, HY-N1475 5mg Nicotiflorin CAS:17650-84-9 Nicotiflorin is a flavonoid glycoside extracted from a traditional Chinese medicine Flos Carthami. Nicotiflorin shows potent antiglycation activity and neuroprotection effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0345 Supplier Medchemexpress LLC Supplier No. HYN14755MG
Medchemexpress, HY-N1475 5mg Nicotiflorin CAS:17650-84-9 Nicotiflorin is a flavonoid glycoside extracted from a traditional Chinese medicine Flos Carthami. Nicotiflorin shows potent antiglycation activity and neuroprotection effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.