Learn More
Cayman Chemical -NIcotIn-d3 10mg
SDP
Supplier:  Cayman Chemical 1825610 MG
Encompass_Preferred
An internal standard for the quantification of (±)-nicotine by GC- or LC-MS
Catalog No. 50-297-9715