Learn More
Cayman Chemical -NIcotIn-d3 25mg
SDP
Catalog No. 502979680
Encompass_Preferred
An internal standard for the quantification of (±)-nicotine by GC- or LC-MS
Catalog No. 50-297-9680 Supplier Cayman Chemical Supplier No. 1825625 MG
An internal standard for the quantification of (±)-nicotine by GC- or LC-MS