Learn More
Cayman Chemical β-Nicotinamide Mononucleotide, Purity: ≥95%, CAS: 1094-61-7, 10 mg
SDP
Catalog No. 502970494
Encompass_Preferred
An intermediate in the biosynthesis of NAD+ that has been used to enhance NAD biosynthesis and glucose-stimulated insulin secretion in Nampt+/- mice and high-fat diet-induced type 2 diabetes mice
Catalog No. 50-297-0494 Supplier Cayman Chemical Supplier No. 1641110 MG
An intermediate in the biosynthesis of NAD+ that has been used to enhance NAD biosynthesis and glucose-stimulated insulin secretion in Nampt+/- mice and high-fat diet-induced type 2 diabetes mice