Learn More
Cayman Chemical NicotinamID MononucleotID 50mg
SDP
Catalog No. 502969948
Encompass_Preferred
An intermediate in the biosynthesis of NAD+ that has been used to enhance NAD biosynthesis and glucose-stimulated insulin secretion in Nampt+/- mice and high-fat diet-induced type 2 diabetes mice
Catalog No. 50-296-9948 Supplier Cayman Chemical Supplier No. 1641150 MG
An intermediate in the biosynthesis of NAD+ that has been used to enhance NAD biosynthesis and glucose-stimulated insulin secretion in Nampt+/- mice and high-fat diet-induced type 2 diabetes mice