Learn More
Cayman Chemical NIcotInamID rIbosIDd4trI 10mg
SDP
Supplier:  Cayman Chemical 3431110 MG
Encompass_Preferred
An internal standard for the quantification of nicotinamide riboside by GC- or LC-MS
Catalog No. 50-295-0789