Learn More
Medchemexpress LLC Nicotinamide 1,N6-ethenoadenine dinucleotide | 38806-38-1 | ≥99.0% | 687.45 | 1 MG
SDP
Catalog No. 5000349157
Encompass_Preferred
Nicotinamide 1,N6-ethenoadenine dinucleotide (ε-NAD) is a fluorescent analog of NAD. This compound is known for its ability to be cleaved by phosphodiesterase I and to bind to bovine liver glutamate dehydrogenase. It functions as a substrate for G-ADP ribosylation of G proteins, a reaction facilitated by bacterial toxins, and is widely used as a fluorescent substrate for the study of ADP ribosylation reactions.
  • Fluorescent analog of NAD
  • Cleaved by phosphodiesterase I (from C. adamanteus venom)
  • Binds to bovine liver glutamate dehydrogenase
  • Serves as a substrate for G-ADP ribosylation of G proteins
  • Used as a fluorescent substrate for studying ADP ribosylation reactions
Catalog No. 50-003-49157 Supplier Medchemexpress LLC Supplier No. HY1375921MG
Nicotinamide 1,N6-ethenoadenine dinucleotide (ε-NAD) is a fluorescent analog of NAD. This compound is known for its ability to be cleaved by phosphodiesterase I and to bind to bovine liver glutamate dehydrogenase. It functions as a substrate for G-ADP ribosylation of G proteins, a reaction facilitated by bacterial toxins, and is widely used as a fluorescent substrate for the study of ADP ribosylation reactions.
  • Fluorescent analog of NAD
  • Cleaved by phosphodiesterase I (from C. adamanteus venom)
  • Binds to bovine liver glutamate dehydrogenase
  • Serves as a substrate for G-ADP ribosylation of G proteins
  • Used as a fluorescent substrate for studying ADP ribosylation reactions