Learn More
Medchemexpress LLC Nicotinamide 1,N6-ethenoadenine dinucleotide | 38806-38-1 | MFCD00063546 | 99.0% | C23H27N7O14P2 | 10MG
SDP
Catalog No. 5000258975
Encompass_Preferred
Nicotinamide 1,N6-ethenoadenine dinucleotide (ε-NAD) is a fluorescent β-NAD analog used as a probe in biochemical and enzymology research. It serves as a fluorescent substrate for studies of nucleotide-protein interactions and ADP-ribosylation reactions. The compound is supplied as a white to off-white solid with high purity and is provided in small research pack sizes with supporting documentation.
Catalog No. 50-002-58975 Supplier Medchemexpress LLC Supplier No. HY13759210MG
Nicotinamide 1,N6-ethenoadenine dinucleotide (ε-NAD) is a fluorescent β-NAD analog used as a probe in biochemical and enzymology research. It serves as a fluorescent substrate for studies of nucleotide-protein interactions and ADP-ribosylation reactions. The compound is supplied as a white to off-white solid with high purity and is provided in small research pack sizes with supporting documentation.