Learn More
Medchemexpress LLC Nicotinamide 1,N6-ethenoadenine dinucleotide (ε-NAD) | 38806-38-1 | MFCD00063546 | 99.0% | 687.45 g/mol | C23H27N7O14P2 | 5 MG
SDP
Catalog No. 5000222431
Encompass_Preferred
Nicotinamide 1,N6-ethenoadenine dinucleotide (ε-NAD) is an etheno-modified nicotinamide adenine dinucleotide supplied as a white to off-white solid for biochemical research. It is used as a fluorescent NAD analogue and substrate in enzymatic and binding studies, including ADP-ribosylation assays.
  • High purity (99%) suitable for enzymatic and analytical applications.
  • Fluorescent etheno modification enables sensitive detection in binding and activity assays.
  • Solid, white to off-white appearance simplifies storage and handling.
  • Available in small batch sizes: 1 MG, 5 MG, 10 MG for screening and assay development.
  • Suitable as a substrate for ADP-ribosylation and nucleotide-binding studies.
Catalog No. 50-002-22431 Supplier Medchemexpress LLC Supplier No. HY1375925MG
Nicotinamide 1,N6-ethenoadenine dinucleotide (ε-NAD) is an etheno-modified nicotinamide adenine dinucleotide supplied as a white to off-white solid for biochemical research. It is used as a fluorescent NAD analogue and substrate in enzymatic and binding studies, including ADP-ribosylation assays.
  • High purity (99%) suitable for enzymatic and analytical applications.
  • Fluorescent etheno modification enables sensitive detection in binding and activity assays.
  • Solid, white to off-white appearance simplifies storage and handling.
  • Available in small batch sizes: 1 MG, 5 MG, 10 MG for screening and assay development.
  • Suitable as a substrate for ADP-ribosylation and nucleotide-binding studies.