Learn More
Medchemexpress LLC HY-B0150 5g , Nicotinamide Niacinamide;Nicotinic acid amide;Vitamin B3 CAS:98-92-0 Purity:98%
SDP
Supplier:  Medchemexpress LLC HYB01505G
Encompass_Preferred
Medchemexpress, HY-B0150 5g Nicotinamide Niacinamide;Nicotinic acid amide;Vitamin B3 CAS:98-92-0 Formula:C6H6N2O PARP-1 Purity:98% Nicotinamide is a water-soluble vitamin, an active component of coenzymes NAD and NADP, and acts as poly (ADP-ribose) polymerase ( PARP ) inhibitor. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2222