Learn More
Cayman Chemical Nicotinamide, Purity: ≥98%, CAS: 98-92-0, 25 mg
SDP
Catalog No. 502952105
Encompass_Preferred
An amide form of niacin; precursor in the synthesis of the metabolic cofactor NAD+; an inhibitor of SIRT1 (IC50 = <50 µM); increases SPT activity and the biosynthesis of ceramide glucosylceramide, sphingomyelin, free fatty acids, and cholesterol in primary human keratinocytes at 10 µM; induces apoptosis in SNU-398, SNU-739, and HepG2 cells at 40 µM; prevents the formation of neoplastic lesions in a diethylnitrosamine-induced mouse model of HCC
Catalog No. 50-295-2105 Supplier Cayman Chemical Supplier No. 1112725 MG
An amide form of niacin; precursor in the synthesis of the metabolic cofactor NAD+; an inhibitor of SIRT1 (IC50 = <50 µM); increases SPT activity and the biosynthesis of ceramide glucosylceramide, sphingomyelin, free fatty acids, and cholesterol in primary human keratinocytes at 10 µM; induces apoptosis in SNU-398, SNU-739, and HepG2 cells at 40 µM; prevents the formation of neoplastic lesions in a diethylnitrosamine-induced mouse model of HCC