Learn More
Selleck Chemical LLC Nicotinamide S1899-10mM/1mL
SDP
Catalog No. 503109763
Encompass_Preferred
Nicotinamide a water-soluble vitamin is an active component of coenzymes NAD and NADP and also act as an inhibitor of sirtuins
Catalog No. 50-310-9763 Supplier Selleck Chemical LLC Supplier No. S189910MM/1ML
Nicotinamide a water-soluble vitamin is an active component of coenzymes NAD and NADP and also act as an inhibitor of sirtuins