Learn More
Selleck Chemical LLC Nicotinamide S1899-5g
SDP
Catalog No. 503110745
Encompass_Preferred
Nicotinamide a water-soluble vitamin is an active component of coenzymes NAD and NADP and also act as an inhibitor of sirtuins
Catalog No. 50-311-0745 Supplier Selleck Chemical LLC Supplier No. S18995G
Nicotinamide a water-soluble vitamin is an active component of coenzymes NAD and NADP and also act as an inhibitor of sirtuins