Learn More
Enzo Life Sciences Nicotinamide (500µl). CAS: 98-92-0
SDP
Supplier:  Enzo Life Sciences BMLKI2830500
Encompass_Preferred
Nicotinamide, the water-soluble amide of nicotinic acid, is a component of the two most important coenzymes - nicotinamide adenine dinucleotide and nicotinamide adenine dinucleotide phosphate. Thus nicotinamide is involved in numerous oxidation-reduction reactions in mammalian biological systems acting as an antioxidant. Formulation: 50 mM Nicotinamide in 50 mM Tris/Cl, pH 8.0, 137 mM NaCl, 2.7 mM KCl, 1 mM MgCl2. Solubility: Soluble in water, 100% ethanol or glycerol. Slightly soluble in chloroform, almost insoluble in ether or acetone. Long Term Storage: -80°C.
Catalog No. 50-201-1112