Learn More
Medchemexpress LLC Nicotinamide | 98-92-0 | 99.9% | 1 G
SDP
Catalog No. 5000406904
Encompass_Preferred
Nicotinamide, also known as Niacinamide, is a form of vitamin B3. It functions as an inhibitor of SIRT1 and SIRT2, increasing cellular NAD+, ATP, and ROS levels. It is involved in various biological processes, including tumor growth inhibition and anti-HBV activity.
  • SIRT2 inhibitor (IC50: 2 μM)
  • SIRT1 inhibitor (IC50: 50-180 μM)
  • Increases cellular NAD+, ATP, and ROS levels
  • Inhibits tumor growth
  • Possesses anti-HBV activity
  • Promotes pancreatic cell differentiation
  • Reduces cell number in melanoma cells
Catalog No. 50-004-06904 Supplier Medchemexpress LLC Supplier No. HYB01501G
Nicotinamide, also known as Niacinamide, is a form of vitamin B3. It functions as an inhibitor of SIRT1 and SIRT2, increasing cellular NAD+, ATP, and ROS levels. It is involved in various biological processes, including tumor growth inhibition and anti-HBV activity.
  • SIRT2 inhibitor (IC50: 2 μM)
  • SIRT1 inhibitor (IC50: 50-180 μM)
  • Increases cellular NAD+, ATP, and ROS levels
  • Inhibits tumor growth
  • Possesses anti-HBV activity
  • Promotes pancreatic cell differentiation
  • Reduces cell number in melanoma cells