Learn More
Cambridge Isotope Laboratories Nicotinamide adenine dinucleotide (NAD⁺), NH₄⁺ salt (ribose-¹³C₅, 98%) CP 96%, 1 mg
Catalog No. NC3307544
Encompass
Molecular Weight- 685.42Chemical Purity- 95%Form- IndividualConcentration- NeatApplication(s)- Biomolecular NMR, Genetic TherapyStorage Temp- Store in freezer (-20°C). Protect from light.
Catalog No. NC3307544 Supplier Cambridge Isotope Laboratories Supplier No. CLM106710.001
Molecular Weight- 685.42Chemical Purity- 95%Form- IndividualConcentration- NeatApplication(s)- Biomolecular NMR, Genetic TherapyStorage Temp- Store in freezer (-20°C). Protect from light.